Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM57 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5141160
This item is not returnable.
View return policy
Description
TMEM57 Polyclonal Antibody for Western Blot
Plays a role in the regulation of neuronal activity.
Specifications
| TMEM57 | |
| Polyclonal | |
| Unconjugated | |
| TMEM57 | |
| 1110007C24Rik; 9230118A01Rik; AI317300; AI606104; brain-specific adapter protein C61; brain-specific adaptor protein C61; C61; MACO1; macoi; Macoilin; Macoilin-1; Tmem57; Transmembrane protein 57 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55219, 66146 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q7TQE6, Q8N5G2 | |
| TMEM57 | |
| Synthetic peptide within Human TMEM57 aa 122-171/664. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction