Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM59 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521065
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521065 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521065 Supplier Thermo Scientific Supplier No. PA521065
Add to Cart
Add to Cart

TMEM59 Polyclonal Antibody

TMEM59 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM59
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TMEM59
Gene Accession No. Q9BXS4
Gene Alias 1110001M20Rik; 3110046P06Rik; AI256529; C1orf8; D4Ertd20e; DCF1; dendritic cell factor 1; HSPC001; Liver membrane-bound protein; MTDCF1; ORF18; PRO195; RGD1310313; RP4-758J24.2; Tdcf1; Thymic dendritic cell-derived factor 1; TMEM59; Transmembrane protein 59; UNQ169; UNQ169/PRO195
Gene Symbols TMEM59
Host Species Rabbit
Immunogen A 16 amino acid peptide near amino terminus of human TMEM59.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9528
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.