Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM59L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542940
Description
Peptide sequence: PAASAPSARD PFAPQLGDTQ NCQLRCRDRD LGPQPSQAGL EGASESPYDR Sequence homology: Human: 100%.
This gene encodes a predicted type-I membrane glycoprotein. The encoded protein may play a role in functioning of the central nervous system.
Specifications
TMEM59L | |
Polyclonal | |
Unconjugated | |
TMEM59L | |
5330410G16Rik; brain-specific membrane-anchored protein; BSMAP; C19orf4; RGD1305557; Tmem59l; transmembrane protein 59 like; transmembrane protein 59-like | |
Rabbit | |
Protein A | |
RUO | |
25789 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UK28 | |
TMEM59L | |
Synthetic peptide directed towards the N-terminal of human TMEM59L (aa 21-70). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction