Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM59L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572688
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572688 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572688 Supplier Invitrogen™ Supplier No. PA572688
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM59L Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a predicted type-I membrane glycoprotein. The encoded protein may play a role in functioning of the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM59L
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TMEM59L
Gene Accession No. Q5HZE8, Q7TNI2, Q9UK28
Gene Alias 5330410G16Rik; brain-specific membrane-anchored protein; BSMAP; C19orf4; RGD1305557; Tmem59l; transmembrane protein 59 like; transmembrane protein 59-like
Gene Symbols TMEM59L
Host Species Rabbit
Immunogen TMEM59L antibody for a 19 amino acid synthetic peptide near carboxy terminus of human TMEM59L. The immunogen is within amino acids 220 - 270 of TMEM59L.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25789, 306349, 67937
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.