Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM59L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532023
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532023 Supplier Invitrogen™ Supplier No. PA532023
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Neuro2A. Predicted reactivity: Pig (88%), Rhesus Monkey (98%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a predicted type-I membrane glycoprotein. The encoded protein may play a role in functioning of the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM59L
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene TMEM59L
Gene Accession No. Q7TNI2, Q9UK28
Gene Alias 5330410G16Rik; brain-specific membrane-anchored protein; BSMAP; C19orf4; RGD1305557; Tmem59l; transmembrane protein 59 like; transmembrane protein 59-like
Gene Symbols TMEM59L
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TMEM59L. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25789, 67937
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.