Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM61 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116922
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116922 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116922 Supplier Invitrogen™ Supplier No. PA5116922
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HT-29, 293T, U-87MG Immunogen sequence: ASIPGPPRWD PYHLSRDLYY LTVESSEKES CRTPKVVDIP TYEEAVSFPV AEGPPTPPAY PTEEALEPSG SRDALLSTQP AWPPPSYESI SLALDAVSAE TTPSATRSCS GLVQTARGGS.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM61
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TMEM61
Gene Accession No. Q8N0U2
Gene Alias TMEM61; Transmembrane protein 61
Gene Symbols TMEM61
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 91-210 of human TMEM61 (NP_8723381).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199964
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.