Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM63A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569093
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569093 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569093 Supplier Invitrogen™ Supplier No. PA569093
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Guinea Pig: 86%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 93%; Yeast: 83%.

Acts as an osmosensitive calcium-permeable cation channel (PubMed:30382938, PubMed:31587869). Mechanosensitive ion channel that converts mechanical stimuli into a flow of ion (PubMed:30382938, PubMed:31587869). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM63A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMEM63A
Gene Accession No. O94886
Gene Alias BC014795; CSC1-like protein 1; KIAA0489; Kiaa0792; RGD1306829; Tmem6; TMEM63A; Tmem64a; transmembrane protein 6; transmembrane protein 63a
Gene Symbols TMEM63A
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human TMEM63A.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9725
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.