Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM63B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541883
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541883 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541883 Supplier Invitrogen™ Supplier No. PA541883
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VRGCEQVEAI EYYTKLEQKL KEDYKREKEK VNEKPLGMAF VTFHNETITA Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.

TMEM63B belongs to the SPO75/TMEM63 family. It is a multi-pass membrane protein. The function of TMEM63B remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM63B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMEM63B
Gene Accession No. Q5T3F8
Gene Alias BC026370; C6orf110; CSC1-like protein 2; RGD1305862; RP3-421H19.2; Tmem63b; Tmem63b-ps1; Transmembrane protein 63B; transmembrane protein 63B, pseudogene 1; transmembrane protein 63B; CSC1-like protein 2
Gene Symbols TMEM63B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TMEM63B.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55362
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.