Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM63B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556094
Description
Immunogen sequence: RTLFINGISK YAESEKIKKH FEEAYPNCTV LEARPCYNVA RLMFLDAERK KAERGKLYFT N Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
TMEM63B belongs to the SPO75/TMEM63 family. It is a multi-pass membrane protein. The function of TMEM63B remains unknown.
Specifications
TMEM63B | |
Polyclonal | |
Unconjugated | |
TMEM63B | |
BC026370; C6orf110; CSC1-like protein 2; RGD1305862; RP3-421H19.2; Tmem63b; Tmem63b-ps1; Transmembrane protein 63B; transmembrane protein 63B, pseudogene 1; transmembrane protein 63B; CSC1-like protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55362 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T3F8 | |
TMEM63B | |
Recombinant protein corresponding to Human TMEM63B. Recombinant protein control fragment (Product #RP-94863). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction