Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM64 Polyclonal Antibody

Supplier: Thermo Scientific PA5112761
Description
Specifications
| TMEM64 | |
| Polyclonal | |
| Unconjugated | |
| TMEM64 | |
| 9630015D15Rik; AI790744; AV300874; CG11367 gene product; Tmem64; Transmembrane protein 64 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 100201, 169200 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q3U145, Q6YI46 | |
| TMEM64 | |
| Recombinant Human Transmembrane protein 64 protein (40-118AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction