Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM65 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555205
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555205 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555205 Supplier Invitrogen™ Supplier No. PA555205
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EALNTAQGAR DFIYSLHSTE RSCLLKELHR FESIAIAQEK LEAPPPT Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 81%.

May play an important role in cardiac development and function. May regulate cardiac conduction and the function of the gap junction protein GJA1. May contribute to the stability and proper localization of GJA1 to cardiac intercalated disk thereby regulating gap junction communication (By similarity). May also play a role in the regulation of mitochondrial respiration and mitochondrial DNA copy number maintenance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM65
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM65
Gene Accession No. Q6PI78
Gene Alias 2610029O13Rik; 4930438D12Rik; TMEM65; Transmembrane protein 65
Gene Symbols TMEM65
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM65. Recombinant protein control fragment (Product #RP-93881).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157378
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.