Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM66 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565267
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565267 Supplier Invitrogen™ Supplier No. PA565267
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YSPPPYSEYPP FSHRYQRFTN SAGPPPPGFK SEFTGPQNTG HGATSGFGSA FT Highest antigen sequence identity to the following orthologs - mouse 75%, rat 70%.

Saraf encodes a protein that is a negative regulator of store-operated Ca(2+) entry (SOCE) involved in protecting cells from Ca(2+) overfilling. In response to cytosolic Ca(2+) elevation after endoplasmic reticulum Ca(2+) refilling, promotes a slow inactivation of STIM (STIM1 or STIM2)-dependent SOCE activity: possibly act by facilitating the de-oligomerization of STIM to efficiently turn off ORAI when the endoplasmic reticulum lumen is filled with the appropriate Ca(2+) levels, and thus preventing the overload of the cell with excessive Ca(2+) ions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM66
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SARAF
Gene Accession No. Q96BY9
Gene Alias 1810045K07Rik; arrestin-E; FOAP-7; HBV XAg-transactivated protein 3; HBV X-transactivated gene 3 protein; HSPC035; NPD003; Protein FOAP-7; PSEC0019; Saraf; SARAF long isoform; SARAF short isoform; SOCE-associated regulatory factor; store-operated calcium entry associated regulatory factor; store-operated calcium entry-associated regulatory factor; testicular secretory protein Li 59; TMEM66; Transmembrane protein 66; UNQ1967/PRO4499; XTP3
Gene Symbols SARAF
Host Species Rabbit
Immunogen Recombinant Human TMEM66. Recombinant protein control fragment (Product #RP-105987).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51669
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.