Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM67 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5141155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5141155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5141155 Supplier Invitrogen™ Supplier No. PA5141155
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM67 Polyclonal Antibody for Western Blot, IHC (P), Flow

The protein encoded by this gene localizes to the primary cilium and to the plasma membrane. The gene functions in centriole migration to the apical membrane and formation of the primary cilium. Multiple transcript variants encoding different isoforms have been found for this gene. Defects in this gene are a cause of Meckel syndrome type 3 (MKS3) and Joubert syndrome type 6 (JBTS6).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM67
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene TMEM67
Gene Accession No. Q5HYA8, Q8BR76
Gene Alias 5330408M12Rik; B230117O07; b2b1163.1Clo; b2b1291.1Clo; JBTS6; meckel syndrome type 3 protein; Meckel syndrome type 3 protein homolog; Meckelin; Mks3; NPHP11; TMEM67; TNEM67; transmembrane protein 67
Gene Symbols TMEM67
Host Species Rabbit
Immunogen Synthetic peptide within human Meckelin aa 20-80.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 329795, 91147
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.