Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM67 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA522095

Catalog No. PIPA522095


Only null left
Add to Cart

Description

Description

Recommended positive controls: 293T, H1299, HeLa, HepG2, Raji. Predicted reactivity: Mouse (82%), Rat (81%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene localizes to the primary cilium and to the plasma membrane. The gene functions in centriole migration to the apical membrane and formation of the primary cilium. Multiple transcript variants encoding different isoforms have been found for this gene. Defects in this gene are a cause of Meckel syndrome type 3 (MKS3) and Joubert syndrome type 6 (JBTS6).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TMEM67
Polyclonal
Unconjugated
TMEM67
5330408M12Rik; B230117O07; b2b1163.1Clo; b2b1291.1Clo; JBTS6; meckel syndrome type 3 protein; Meckel syndrome type 3 protein homolog; Meckelin; Mks3; NPHP11; TMEM67; TNEM67; transmembrane protein 67
Rabbit
Antigen affinity chromatography
RUO
91147
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q5HYA8
TMEM67
Recombinant fragment corresponding to a region within amino acids 231 and 510 of meckelin isoform 1 (Uniprot ID#Q5HYA8).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.