Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM70 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554717
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554717 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554717 Supplier Invitrogen™ Supplier No. PA554717
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KGYVIRLYHE ATTDTYKAIT YNAMLAETST VFHQNDVKIP DAKHVFTTFY AKTKSLLVNP VLFPNREDYI HLMGYDKEEF ILYMEETSEE KRHKDDK Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 71%.

TMEM70 is a recently identified mitochondrial protein that is thought to play a role in the biogenesis of the ATP synthase in higher eukaryotes. Mutations in this gene result in early neonatal onset of hypotonia, hypertrophic cardiomyopathy, lactic acidos and 3-methylglutaconic aciduria (3-MGC-uria), and usually cause death within the first six weeks of life, although some patients survive much longer. Little is known of the role of TMEM70, but it is conserved across multicellular eukaryotic organisms. It contains a conserved DUF1301 domain and two putative transmembrane regions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM70
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM70
Gene Accession No. Q9BUB7
Gene Alias 1110020A09Rik; 2210416J16Rik; MC5DN2; RGD1566224; TMEM70; transmembrane protein 70; transmembrane protein 70, mitochondrial
Gene Symbols TMEM70
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM70. Recombinant protein control fragment (Product #RP-91012).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54968
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.