Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM71 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567117
Description
Immunogen sequence: MSTPVASSSRL EREYAGELSP TCIFPSFTCD SLDGYHSFEC GSIDPLTGSH YTCRRSPRLL TNGYYIWTED Highest antigen sequence identity to the following orthologs - mouse 79%, rat 69%.
TMEM71 is a protein coding gene.
Specifications
TMEM71 | |
Polyclonal | |
Unconjugated | |
TMEM71 | |
TMEM71; Transmembrane protein 71 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
137835 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6P5X7 | |
TMEM71 | |
Recombinant Human TMEM71. Recombinant protein control fragment (Product #RP-107761). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction