Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM72 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558966
Description
Immunogen sequence: KRKKRKAAPE VLASPEQYTD PSSSAVSTTG SGDTEQTYTF HGALKEGPSS LFIHMKSILK GTKKPSA Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 81%.

Specifications
TMEM72 | |
Polyclonal | |
Unconjugated | |
TMEM72 | |
C10orf127; Kidney-specific secretory protein of 37 kDa; KSP37; TMEM72; Transmembrane protein 72 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
643236 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A0PK05 | |
TMEM72 | |
Recombinant protein corresponding to Human TMEM72. Recombinant protein control fragment (Product #RP-97190). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction