Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM74 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587853
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587853 Supplier Invitrogen™ Supplier No. PA587853
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MELHYLAKKS NQADLCDARD WSSRGLPGDQ ADTAATRAAL CCQKQCASTP RATEMEGSKL SSSPASPSSS LQNSTLQPDA FPPGLLHSGN NQITAERKVC NCCSQELETS FTYVDKNINL EQRNRSSPSA KGHNHPGELG WENPNEWSQE AAISLISEEE DDTSSEATSS GKSIDYGFIS; Positive Samples: DU145; Cellular Location: Cytoplasmic vesicle, Lysosome membrane, Multi-pass membrane protein, autophagosome membrane.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM74
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TMEM74
Gene Accession No. Q96NL1
Gene Alias AA549547; B230382K22Rik; NET36; RGD1566279; TMEM74; Transmembrane protein 74
Gene Symbols TMEM74
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human TMEM74 (NP_6945601).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157753
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.