Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM80 Antibody (OTI3A6), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274546AF700
Description
TMEM80 Monoclonal specifically detects TMEM80 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
TMEM80 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TMEM80 | |
Full length recombinant protein of human TMEM80 (NP_777600) produced in HEK293T cells. | |
0.1 mL | |
283232.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI3A6 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
FLJ38216, FLJ55823, transmembrane protein 80 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction