Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM80 Antibody (OTI3A6), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274546MFV450
Description
TMEM80 Monoclonal antibody specifically detects TMEM80 in Human samples. It is validated for Flow CytometrySpecifications
TMEM80 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
283232 | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
OTI3A6 | |
Flow Cytometry | |
FLJ38216, FLJ55823, transmembrane protein 80 | |
Full length human recombinant protein of human TMEM80 (NP_777600) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction