Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM80 Monoclonal Antibody (OTI2B7)
GREENER_CHOICE

Catalog No. PIMA525181
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525181 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525181 Supplier Invitrogen™ Supplier No. MA525181
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TMEM80 Monoclonal Antibody for Western Blot, ICC/IF, Flow

TMEM80 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM80
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI2B7
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TMEM80
Gene Accession No. Q96HE8
Gene Alias 5530601I19Rik; Tmem80; Transmembrane protein 80
Gene Symbols TMEM80
Host Species Mouse
Immunogen Full length human recombinant protein of TMEM80 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283232
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.