Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM81 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565728
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565728 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565728 Supplier Invitrogen™ Supplier No. PA565728
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLRLVKRLYFG LRVLPPNLVN LNFHQSLTED QKLIDEGLEV NLDSYSKPHH PKWK Highest antigen sequence identity to the following orthologs - mouse 80%, rat 76%.

TMEM81 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM81
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM81
Gene Accession No. Q6P7N7
Gene Alias HC3107; KVLA2788; TMEM81; Transmembrane protein 81; UNQ2788; UNQ2788/PRO7178
Gene Symbols TMEM81
Host Species Rabbit
Immunogen Recombinant Human TMEM81. Recombinant protein control fragment (Product #RP-104948).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388730
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.