Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM88 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521164
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521164 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521164 Supplier Invitrogen™ Supplier No. PA521164
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human brain tissue lysate. PA5-21164 can be used with blocking peptide PEP-1278.

Transmembrane protein 88 (TMEM88) is a two-transmembrane-type protein whose C-terminal tail has been shown to bind the PDZ domain of Dishevelled (Dvl), one of the key components in Wnt signaling pathways. TMEM88 attentuated the Wnt/beta-catenin signaling induced by Wnt-1 ligand in a dose-dependent manner and knockdown of TMEM88 by RNAi increased Wnt activity, suggesting that TMEM88 plays a role in regulating Wnt signaling in a context-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM88
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TMEM88
Gene Accession No. Q6PEY1, Q9D0N8
Gene Alias 2600017H02Rik; RGD1561781; TMEM88; TMEM88A; Transmembrane protein 88
Gene Symbols TMEM88
Host Species Rabbit
Immunogen A 19 amino acid peptide near the carboxy terminus of human TMEM88.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 497936, 67020, 92162
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.