Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM8A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566706
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566706 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566706 Supplier Invitrogen™ Supplier No. PA566706
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PFLGFNTSLNC TTAFFQGYPL SLSAWSRRAN LIIPYPETDN WYLSLQLMCP ENAEDCEQAV VHVETTLYLV PC Highest antigen sequence identity to the following orthologs - mouse 81%, rat 82%.

Involved in the lipid remodeling steps of GPI-anchor maturation. Lipid remodeling steps consist in the generation of 2 saturated fatty chains at the sn-2 position of GPI-anchor proteins (GPI-AP). Has phospholipase A2 activity that removes an acyl-chain at the sn-2 position of GPI-anchors during the remodeling of GPI. Required for the shedding of the GPI-AP TDGF1, but not CFC1, at the cell surface. Shedding of TDGF1 modulates Nodal signaling by allowing soluble TDGF1 to act as a Nodal coreceptor on other cells (PubMed:27881714). Also indirectly involved in the translocation of RAC1 from the cytosol to the plasma membrane by maintaining the steady state amount of CAV1-enriched plasma membrane subdomains, stabilizing RAC1 at the plasma membrane (PubMed:27835684). In contrast to myomaker (TMEM8C), has no fusogenic activity (PubMed:26858401). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM8A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM8A
Gene Accession No. Q9HCN3
Gene Alias five-span transmembrane protein M83; GPI processing phospholipase A2; GPI-PLA2; M83; PGAP6; Post-GPI attachment to proteins factor 6; Protein M83; TMEM6; TMEM8; TMEM8A; Transmembrane protein 6; Transmembrane protein 8; transmembrane protein 8 (five membrane-spanning domains); transmembrane protein 8A
Gene Symbols TMEM8A
Host Species Rabbit
Immunogen Recombinant Human TMEM8A. Recombinant protein control fragment (Product #RP-107355).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58986
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.