Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM8A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562365
Description
Immunogen sequence: TCAYVFQPEL LVTRVVEISI MEPDVPLPQT LLSHPSYLKV FVPDYTRELL LELRDCVSNG SLGCPVRLTV GPVTLPSNFQ KVL Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 75%.
Involved in the lipid remodeling steps of GPI-anchor maturation. Lipid remodeling steps consist in the generation of 2 saturated fatty chains at the sn-2 position of GPI-anchor proteins (GPI-AP). Has phospholipase A2 activity that removes an acyl-chain at the sn-2 position of GPI-anchors during the remodeling of GPI. Required for the shedding of the GPI-AP TDGF1, but not CFC1, at the cell surface. Shedding of TDGF1 modulates Nodal signaling by allowing soluble TDGF1 to act as a Nodal coreceptor on other cells (PubMed:27881714). Also indirectly involved in the translocation of RAC1 from the cytosol to the plasma membrane by maintaining the steady state amount of CAV1-enriched plasma membrane subdomains, stabilizing RAC1 at the plasma membrane (PubMed:27835684). In contrast to myomaker (TMEM8C), has no fusogenic activity (PubMed:26858401). [UniProt].
Specifications
TMEM8A | |
Polyclonal | |
Unconjugated | |
TMEM8A | |
five-span transmembrane protein M83; GPI processing phospholipase A2; GPI-PLA2; M83; PGAP6; Post-GPI attachment to proteins factor 6; Protein M83; TMEM6; TMEM8; TMEM8A; Transmembrane protein 6; Transmembrane protein 8; transmembrane protein 8 (five membrane-spanning domains); transmembrane protein 8A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
58986 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HCN3 | |
TMEM8A | |
Recombinant protein corresponding to Human TMEM8A. Recombinant protein control fragment (Product #RP-101281). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction