Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM8B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542186
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542186 Supplier Invitrogen™ Supplier No. PA542186
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: THNYLYAACE CKAGWRGWGC TDSADALTYG FQLLSTLLLC LSNLMFLPPV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

The down-regulation of C9orf127 may be closely associated with tumorigenesis and metastasis of colorectal carcinoma. However, it may not contribute to the development and progression of gastric carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM8B
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMEM8B
Gene Accession No. A6NDV4
Gene Alias 4930500O05Rik; C9orf127; NAG5; NAG-5; nasopharyngeal carcinoma expressed 6; nasopharyngeal carcinoma related protein; nasopharyngeal carcinoma-associated gene 6 protein; NGX6; Protein NAG-5; Protein NGX6; RGD1310012; Tmem8b; transmembrane protein 8A-like; transmembrane protein 8B
Gene Symbols TMEM8B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human C9orf127.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51754
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.