Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100207
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100207 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100207 Supplier Thermo Scientific Supplier No. PA5100207
Add to Cart
Add to Cart

TMEM9 Polyclonal Antibody

TMEM9 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM9
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TMEM9
Gene Accession No. Q9CR23, Q9P0T7
Gene Alias 1500015G18Rik; AW545782; DERM4; dermal papilla-derived protein 4; DERP4; fi33g11; HSPC186; PSEC0012; TMEM9; TMEM9A; Transmembrane protein 9; UNQ631/PRO1248; wu:fi33g11; zgc:55432
Gene Symbols TMEM9
Host Species Rabbit
Immunogen A synthesized peptide derived from human TMEM9(Accession Q9P0T7), corresponding to amino acid residues L14-V64.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 252839, 289046, 66241
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.