Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM91 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112765
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112765 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112765 Supplier Invitrogen™ Supplier No. PA5112765
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM91 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM91
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TMEM91
Gene Accession No. Q6ZNR0
Gene Alias dispanin subfamily C member 3; DSPC3; IFITMD6; interferon induced transmembrane protein domain containing 6; TMEM91; Transmembrane protein 91
Gene Symbols TMEM91
Host Species Rabbit
Immunogen Recombinant Human Transmembrane protein 91 protein (1-97AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 641649
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.