Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM95 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144113
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144113 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144113 Supplier Invitrogen™ Supplier No. PA5144113
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM95 Polyclonal Antibody for ELISA, ICC/IF, IHC (P)

Sperm protein required for fusion of sperm with the egg membrane during fertilization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM95
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene TMEM95
Gene Accession No. Q3KNT9
Gene Alias TMEM95; Transmembrane protein 95; UNQ9390/PRO34281
Gene Symbols TMEM95
Host Species Rabbit
Immunogen Recombinant Human Transmembrane protein 95 protein (17-145AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339168
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.