Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM97 Antibody, FITC, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP130436F

Catalog No. NBP130436F


Only null left
Add to Cart

Description

Description

TMEM97 Polyclonal specifically detects TMEM97 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

TMEM97
Polyclonal
Immunocytochemistry/Immunofluorescence
Q5BJF2
TMEM97
Synthetic peptide made to an internal portion of the human TMEM97 protein (within residues 105-155. [Swiss-Prot: Q5BJF2]
Affinity Purified
RUO
Primary
Human. Predicted to react with monkey based on 100% sequence homology.
Store at 4C in the dark.
Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
MAC30Protein MAC30, transmembrane protein 97
Rabbit
24 kDa
0.1 mL
Cholesterol Metabolism, Cytokine Research, Ovarian Carcinoma Cell Markers
27346
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only