Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM97 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140288
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140288 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140288 Supplier Invitrogen™ Supplier No. PA5140288
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DLQAVLPREL YPVEFRNLLK WYAKEFKDPL LQEPPAWFK Highest antigen sequence indentity to the following orthologs: Mouse - 82%, Rat - 79%.

TMEM97, or MAC30, is a conserved transmembrane protein that helps regulate cellular cholesterol levels through colocalization with NPC1. TMEM97 has also been linked to several cancers including breast, pancreatic, colorectal and ovarian cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM97
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM97
Gene Accession No. Q5BJF2
Gene Alias 1810014L12Rik; AI115531; AL022956; D11Bhm182e; MAC30; meningioma-associated protein 30; S2R; Sigma intracellular receptor 2; Sigma-2 receptor; Tmem97; Transmembrane protein 97
Gene Symbols TMEM97
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM97. Recombinant protein control fragment (Product #RP-109584).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27346
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.