Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM98 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559192
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559192 Supplier Invitrogen™ Supplier No. PA559192
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CRQRYCRPRD LLQRYDSKPI VDLIGAMETQ SEPSELELDD VVITNPHIEA ILENEDWIED ASGLMSHCIA ILKICHTLTE KLVAMTMGSG AKMKTSASVS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene encodes a transmembrane protein. A missense mutation in this gene result in Nanophthalmos 4 (NNO4). Alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM98
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM98
Gene Accession No. Q9Y2Y6
Gene Alias 6530411B15Rik; AI463522; Protein TADA1; TADA1; TMEM98; Transmembrane protein 98; UNQ536/PRO1079
Gene Symbols TMEM98
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM98. Recombinant protein control fragment (Product #RP-89947).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26022
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.