Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEPAI Mouse anti-Human, Clone: 9F10, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS1905
Description
This mouse monoclonal Anti-TMEPAI, Antibody, clone 9F10 Immunoprecipitation, and Western blotting for the detection of TMEPAI.
Specifications
TMEPAI | |
Monoclonal | |
Please refer to lot specific datasheet. | |
PMEPA1,STAG1,TMEPAI | |
GST-tagged recombinant fragment corresponding to 39 amino acids from the C-terminal region of human TMEPAI. | |
100 μg | |
Cell Structure | |
NP_064567 | |
Human | |
Purified |
Western Blot, Immunoprecipitation, Immunocytochemistry | |
9F10 | |
Unconjugated | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone 9F10 detects TMEPAI in COS-7 cells overexpressing TEMPAI. It targets an epitope of 39 amino acids from the C-terminal region in isoform 1. Also shows reactivity with isoform 2 that has 100% sequence homology in this epitope region. | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction