Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEPAI, Mouse anti-Human, Clone: PMEPA1/2698, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279758100UG
Description
TMEPAI Monoclonal specifically detects TMEPAI in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
TMEPAI | |
Monoclonal | |
0.2 mg/mL | |
Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
prostate transmembrane protein, androgen induced 1, Solid tumor-associated 1 protein, STAG1prostate androgen induced RNA, transmembrane prostate androgen-induced protein | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
PMEPA1/2698 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
PMEPA1 | |
Recombinant full-length human TMEPAI protein | |
100 μg | |
Signal Transduction | |
56937 | |
Store at 4°C. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction