Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531184
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531184 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531184 Supplier Invitrogen™ Supplier No. PA531184
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG, mouse testis. Predicted reactivity: Mouse (86%), Rat (83%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Adapter protein that may provide indirect link between the endocytic membrane traffic and the actin assembly machinery. May regulate the formation of clathrin-coated vesicles. Isoform 1 could be involved in brain-specific synaptic vesicle recycling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMF1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene TMF1
Gene Accession No. B9EKI3, P82094
Gene Alias 7030402D04Rik; Androgen receptor coactivator 160 kDa protein; androgen receptor-associated protein of 160 kDa; ARA160; Gm153; TATA element modulatory factor; TATA element modulatory factor 1; TMF; TMF1; Tmf1
Gene Symbols TMF1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TMF1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114206, 232286, 7110
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.