Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMIE Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558330
Description
Immunogen sequence: RTRKEIEARY LQRKAAKMYT DKLETVPPLN ELTEVPGEDK KKKK Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
This gene encodes a transmembrane inner ear protein. Studies in mouse suggest that this gene is required for normal postnatal maturation of sensory hair cells in the cochlea, including correct development of stereocilia bundles. This gene is one of multiple genes responsible for recessive non-syndromic deafness (DFNB), also known as autosomal recessive nonsyndromic hearing loss (ARNSHL), the most common form of congenitally acquired inherited hearing impairment.
Specifications
TMIE | |
Polyclonal | |
Unconjugated | |
TMIE | |
5131400L21Rik; DFNB6; Mm.87012; spinner; sr; Tmie; transmembrane inner ear; transmembrane inner ear expressed protein; transmembrane inner ear protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
259236 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NEW7 | |
TMIE | |
Recombinant protein corresponding to Human TMIE. Recombinant protein control fragment (Product #RP-96584). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction