Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMIGD1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300307AF750
Description
TMIGD1 Polyclonal specifically detects TMIGD1 in Human samples. It is validated for ELISA.Specifications
TMIGD1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
388364 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 750 | |
AWKS9372, TMIGD, transmembrane and immunoglobulin domain containing 1, transmembrane and immunoglobulin domain-containing protein 1, UNQ9372 | |
Produced in rabbits immunized with purified, recombinant TMIGD1 (NP_996663.1; Met1-Pro220). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction