Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMIGD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA538057
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA538057 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA538057 Supplier Invitrogen™ Supplier No. PA538057
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A recommended positive control is rat liver tissue lysate. At least two isoforms of TMIGD1 are known to exist; this antibody will detect both isoforms.

TMIGD1 (transmembrane and immunoglobulin domain containing 1) is a 262 amino acid single-pass membrane protein that contains two Ig-like C2-type (immunoglobulin-like) domains. It is a nonsecreted, glycosylated protein located in the cytoplasm and cell membrane. TMIGD1 might be a possible cell-differentiation marker in the intestine, whose expression is lost during cellular transformation and promote progression of tumorigenesis in the intestinal epithelium. TMIGD1 has also been postulated to play a role in cell adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMIGD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TMIGD1
Gene Accession No. Q6UXZ0, Q9D7L8
Gene Alias 2010002A20Rik; AWKS9372; RGD1307444; TMIGD; Tmigd1; transmembrane and immunoglobulin domain containing 1; transmembrane and immunoglobulin domain-containing protein 1; UNQ9372; UNQ9372/PRO34164
Gene Symbols TMIGD1
Host Species Rabbit
Immunogen 18 amino acid peptide near the center of human TMIGD1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363654, 388364, 66601
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.