Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMIGD1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989060
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9060 Supplier Bioss Supplier No. BS9427R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMIGD1 (transmembrane and immunoglobulin domain containing 1) is a 262 amino acid single-pass membrane protein that contains two Ig-like C2-type (immunoglobulin-like) domains. It is a nonsecreted, glycosylated protein located in the cytoplasm and cell membrane. TMIGD1 might be a possible cell-differentiation marker in the intestine, whose expression is lost during cellular transformation and promote progression of tumorigenesis in the intestinal epithelium. TMIGD1 has also been postulated to play a role in cell adhesion.

Specifications

Antigen TMIGD1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TMIGD1
Gene Accession No. Q6UXZ0
Gene Alias 2010002A20Rik; AWKS9372; RGD1307444; Tmigd; Tmigd1; transmembrane and immunoglobulin domain containing 1; Transmembrane and immunoglobulin domain-containing protein 1; UNQ9372; UNQ9372/PRO34164
Gene Symbols TMIGD1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TMIGD1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388364
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.