Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMIGD2 Antibody (953728) [PerCP], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8316C
Description
TMIGD2 Monoclonal antibody specifically detects TMIGD2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
TMIGD2 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
126259 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, Immunohistochemistry, CyTOF | |
953728 | |
Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
CD28H, IGPR1, IGPR-1, MGC23244, transmembrane and immunoglobulin domain containing 2, transmembrane and immunoglobulin domain-containing protein 2 | |
HEK293 human embryonic kidney cell line-derived human TMIGD2/CD28H, Met1-Gly150, Accession # Q96BF3 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction