Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMIGD2 Antibody (953730), Alexa Fluor™ 488, R&D Systems™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB83161AF488
Description
TMIGD2 Monoclonal specifically detects TMIGD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Product Image.Specifications
TMIGD2 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TMIGD2 | |
Partial recombinant human CD28H protein (amino acids 23-150) [UniProt Q96BF3] | |
100 μL | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
953730 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CD28H, IGPR1, IGPR-1, MGC23244, transmembrane and immunoglobulin domain containing 2, transmembrane and immunoglobulin domain-containing protein 2 | |
Mouse | |
Protein A or G purified | |
RUO | |
126259 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction