Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMIGD2 Monoclonal Antibody (7A10)
GREENER_CHOICE

Catalog No. PIMA549141
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549141 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549141 Supplier Invitrogen™ Supplier No. MA549141
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TMIGD2 Monoclonal Antibody for ICC/IF, IHC (P)

TMIGD2 (transmembrane and immunoglobulin domain containing 1), also known as immunoglobulin-containing and proline-rich receptor 1 (IGPR1), is novel adhesion molecule that is expressed in multiple tissues, primarily in cells of epithelium and endothelium origins. TMIGD2 is thought to be involved in angiogenesis and regulates cellular morphology, homophilic cell aggregation, and cell-cell interaction. TMIGD2 activity also modulates actin stress fiber formation and focal adhesion and reduces cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMIGD2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone 7A10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene TMIGD2
Gene Accession No. Q96BF3
Gene Alias CD28 homolog; CD28 homologue; CD28H; IGPR1; IGPR-1; Immunoglobulin and proline-rich receptor 1; immunoglobulin-containing and proline-rich receptor 1; immunoglobulin-containing and proline-rich receptor-1; TMIGD2; transmembrane and immunoglobulin domain containing 2; transmembrane and immunoglobulin domain-containing protein 2; transmembrane and immunoglobulin domain-containing protein 2 variant 2; transmembrane and immunoglobulin domain-containing protein 2 variant 3; UNQ3059/PRO9879
Gene Symbols TMIGD2
Host Species Mouse
Immunogen The extracellular domain of human TMIGD2.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126259
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.