Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMLHE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521461 Supplier Invitrogen™ Supplier No. PA521461
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: H1299, HeLaS3, HepG2. Predicted reactivity: Mouse (95%), Rat (92%), Chicken (91%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes the protein trimethyllysine dioxygenase which is the first enzyme in the carnitine biosynthesis pathway. Carnitine play an essential role in the transport of activated fatty acids across the inner mitochondrial membrane. The encoded protein converts trimethyllysine into hydroxytrimethyllysine. A pseudogene of this gene is found on chromosome X. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMLHE
Applications Western Blot
Classification Polyclonal
Concentration 6.68 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Tmlhe
Gene Accession No. Q9NVH6
Gene Alias AUTSX6; Bbox2; butyrobetaine (gamma), 2-oxoglutarate dioxygenase (gamma-butyrobetaine hydroxylase) 2; butyrobetaine (gamma), 2-oxoglutarate dioxygenase 2 (gamma-butyrobetaine hydroxylase); D430017M14Rik; epsilon-trimethyllysine 2-oxoglutarate dioxygenase; Epsilon-trimethyllysine hydroxylase; si:ch211-145n14.3; TML dioxygenase; TML hydroxylase; TML-alpha-ketoglutarate dioxygenase; TMLD; Tmlh; TMLHE; TMLHED; Trimethyllysine dioxygenase, mitochondrial; trimethyllysine hydroxylase, epsilon; XAP130
Gene Symbols Tmlhe
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TMLHE. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55217
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.