Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMLHE Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556983
Description
Immunogen sequence: AYTKLALDRH TDTTYFQEPC GIQVFHCLKH EGTGGRTLLV DGFYAAEQVL QKAPEEFELL SKVPLKHEYI EDVGECHNHM IGIGPV Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 91%.
This gene encodes the protein trimethyllysine dioxygenase which is the first enzyme in the carnitine biosynthesis pathway. Carnitine play an essential role in the transport of activated fatty acids across the inner mitochondrial membrane. The encoded protein converts trimethyllysine into hydroxytrimethyllysine. A pseudogene of this gene is found on chromosome X. Alternate splicing results in multiple transcript variants.
Specifications
| TMLHE | |
| Polyclonal | |
| Unconjugated | |
| Tmlhe | |
| AUTSX6; Bbox2; butyrobetaine (gamma), 2-oxoglutarate dioxygenase (gamma-butyrobetaine hydroxylase) 2; butyrobetaine (gamma), 2-oxoglutarate dioxygenase 2 (gamma-butyrobetaine hydroxylase); D430017M14Rik; epsilon-trimethyllysine 2-oxoglutarate dioxygenase; Epsilon-trimethyllysine hydroxylase; si:ch211-145n14.3; TML dioxygenase; TML hydroxylase; TML-alpha-ketoglutarate dioxygenase; TMLD; Tmlh; TMLHE; TMLHED; Trimethyllysine dioxygenase, mitochondrial; trimethyllysine hydroxylase, epsilon; XAP130 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55217 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NVH6 | |
| Tmlhe | |
| Recombinant protein corresponding to Human TMLHE. Recombinant protein control fragment (Product #RP-95470). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction