Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMLHE Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | TMLHE |
---|---|
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
TMLHE Polyclonal specifically detects TMLHE in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
TMLHE | |
Polyclonal | |
Purified | |
RUO | |
Human | |
BBOX2TML-alpha-ketoglutarate dioxygenase, butyrobetaine (gamma), 2-oxoglutarate dioxygenase (gamma-butyrobetainehydroxylase) 2, EC 1.14.11.8, Epsilon-trimethyllysine 2-oxoglutarate dioxygenase, Epsilon-trimethyllysine hydroxylase, FLJ10727, TML dioxygenase, TMLD, TMLHTML hydroxylase, trimethyllysine dioxygenase, mitochondrial, trimethyllysine hydroxylase, epsilon, XAP130 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 167-376 of human TMLHE (NP_001171726.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
DNA Repair, Mismatch Repair | |
PBS with 50% glycerol, pH7.3. | |
55217 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title