Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMM35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561774
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561774 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561774 Supplier Invitrogen™ Supplier No. PA561774
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRLSKDAYSE MKRAYKSYVR ALPLLKKMGI NSILLRKSIG ALEVACGIVM TLVPGRPKDV Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

A soluble peptide released by shedding may interact with NGFR and modulate neurite outgrowth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMM35
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM35A
Gene Accession No. Q53FP2
Gene Alias 9030603L14Rik; AI841526; Peroxisomal membrane protein 52; PMP52; TMEM35; TMEM35A; transmembrane protein 35; transmembrane protein 35A
Gene Symbols TMEM35A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMM35. Recombinant protein control fragment (Product #RP-92201).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59353
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.