Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMOD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | TMOD1 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6059
![]() ![]() |
Proteintech Group Inc
101451AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
TMOD1 gene encodes a member of the tropomodulin family. The encoded protein is an actin-capping protein that regulates tropomyosin by binding to its N-terminus, inhibiting depolymerization and elongation of the pointed end of actin filaments and thereby influencing the structure of the erythrocyte membrane skeleton. Multiple transcript variants encoding the same protein have been found for this gene.Specifications
TMOD1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P28289, P49813, P70567 | |
21916, 25566, 7111 | |
TMOD1 Fusion Protein Ag0192 | |
Primary | |
-20°C | |
TMOD1 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
D9S57E, E Tmod, Erythrocyte tropomodulin, ETMOD, TMOD, TMOD1, tropomodulin 1 | |
Tmod1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title