Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMOD2 Recombinant Rabbit Monoclonal Antibody (5E7G5)
GREENER_CHOICE

Catalog No. PIMA542737
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542737 Supplier Invitrogen™ Supplier No. MA542737
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: SH-SY5Y, U-251MG, Mouse testis, Mouse brain, Rat brain. Immunogen sequence: MALPFQKELE KYKNIDEDEL LGKLSEEELK QLENVLDDLD PESAMLPAGF RQKDQTQKAA TGPFDREHLL MYLEKEALEQ KDREDFVPFT GEKKGRVFIP.

This gene encodes a neuronal-specific member of the tropomodulin family of actin-regulatory proteins. The encoded protein caps the pointed end of actin filaments preventing both elongation and depolymerization. The capping activity of this protein is dependent on its association with tropomyosin. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMOD2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 5E7G5
Concentration 0.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Tmod2
Gene Accession No. P70566, Q9JKK7, Q9NZR1
Gene Alias neural tropomodulin; neuronal tropomodulin; NTMOD; N-TMOD; TMOD2; tropomodulin 2; tropomodulin 2 (neuronal); tropomodulin-2
Gene Symbols Tmod2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TMOD2 (Q9NZR1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29767, 50876, 58814
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.