Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMOD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544270
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544270 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544270 Supplier Invitrogen™ Supplier No. PA544270
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LDDLDPENAL LPAGFRQKNQ TSKSTTGPFD REHLLSYLEK EALEHKDRED Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%.

Blocks the elongation and depolymerization of the actin filaments at the pointed end. The Tmod/TM complex contributes to the formation of the short actin protofilament, which in turn defines the geometry of the membrane skeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMOD3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Tmod3
Gene Accession No. Q9NYL9
Gene Alias actin filament pointed-end capping protein; Tmod3; tropomodulin 3; tropomodulin 3 (ubiquitous); tropomodulin 3; ubiquitous tropomodulin; tropomodulin-3; Tropomodulin-3-like protein; Ubiquitous tropomodulin; ubiquitous tropomodulin U-Tmod; Unknown (protein for MGC:127550); UTMOD; U-Tmod
Gene Symbols Tmod3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TMOD3 (aa 36-85).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29766
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.