Learn More
Invitrogen™ TMOD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551645
Description
Immunogen sequence: LEKEALEHKD REDYVPYTGE KKGKIFIPKQ KPVQTFTEEK VSLDPELEEA LTSASDTELC DLAAILGMHN LITNTKFCNI MGSSNGVDQE HFSNVVKGEK ILPVFDEPPN PTNVEESLKR TKENDAHLVE VNLNNIKNIP IPTLKD Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.
Blocks the elongation and depolymerization of the actin filaments at the pointed end. The Tmod/TM complex contributes to the formation of the short actin protofilament, which in turn defines the geometry of the membrane skeleton.
Specifications
TMOD3 | |
Polyclonal | |
Unconjugated | |
Tmod3 | |
actin filament pointed-end capping protein; Tmod3; tropomodulin 3; tropomodulin 3 (ubiquitous); tropomodulin 3; ubiquitous tropomodulin; tropomodulin-3; Tropomodulin-3-like protein; Ubiquitous tropomodulin; ubiquitous tropomodulin U-Tmod; Unknown (protein for MGC:127550); UTMOD; U-Tmod | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29766, 300838, 50875 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9JHJ0, Q9NYL9 | |
Tmod3 | |
Recombinant protein corresponding to Human TMOD3. Recombinant protein control fragment (Product #RP-101022). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.